Assay Details
Target Gene Details
Entrez Gene ID: | 380768 |
Gene Name: | coiled-coil domain containing 177 |
Gene Aliases: |
Gm1568 |
Location: |
Chr.12:80755447-80760715 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc177 | NM_001008423.2 | 1 | 51 | NP_001008423.2 |
BC067044.1 | AAH67044.1 | |||
BY727768.1 | 1 | 56 |