Assay Details
Target Gene Details
Entrez Gene ID: | 17940 |
Gene Name: | NLR family, apoptosis inhibitory protein 1 |
Gene Aliases: |
AV364616, Birc1a, D13Lsd1, Naip, Naip-rs1 |
Location: |
Chr.13:100407764-100452864 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Naip1 | NM_008670.2 | 11 | 3036 | NP_032696.2 |
AF007769.1 | 10 | 3133 | AAB69223.1 | |
AF135491.1 | 11 | 3036 | AAD56763.1 | |
BC132413.1 | 10 | 3083 |
More Information
Set Membership: |
Intragenic Exonic |