Assay Details
Target Gene Details
Entrez Gene ID: | 18191 |
Gene Name: | neurexin III |
Gene Aliases: |
- |
Location: |
Chr.12:88722865-90334935 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrxn3 | NM_001198587.3 | 7 | 2655 | NP_001185516.2 |
NM_172544.3 | 6 | 1545 | NP_766132.2 | |
AK032126.1 | 6 | 1537 | BAC27716.1 | |
AK078216.1 | 8 | 2967 | ||
AK082055.1 | 8 | 2967 | ||
AK129205.1 | 5 | 1418 | BAC98015.2 | |
BC060719.1 | 8 | 2230 | AAH60719.1 |