Assay Details
Target Gene Details
Entrez Gene ID: | 18191 |
Gene Name: | neurexin III |
Gene Aliases: |
- |
Location: |
Chr.12:88722865-90334935 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrxn3 | NM_001198587.3 | 20 | 5565 | NP_001185516.2 |
NM_001252074.2 | 7 | 2789 | NP_001239003.1 | |
NM_172544.3 | 20 | 4161 | NP_766132.2 | |
AK032126.1 | 20 | 4153 | BAC27716.1 | |
AK034255.1 | 2 | 1791 | BAC28651.1 | |
AK034555.1 | ||||
AK036342.1 | 1 | 659 | BAE20501.1 | |
AK042730.1 | 7 | 2791 | BAC31344.1 | |
AK129205.1 | 19 | 5283 | BAC98015.2 | |
BC060719.1 | 21 | 5140 | AAH60719.1 |
More Information
Set Membership: |
Intragenic Exonic |