Assay Details
Target Gene Details
Entrez Gene ID: | 238057 |
Gene Name: | growth differentiation factor 7 |
Gene Aliases: |
BMP12 |
Location: |
Chr.12:8291018-8302088 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gdf7 | NM_013527.1 | 1 | 259 | NP_038555.1 |
AF525752.1 | 1 | 259 | AAP97721.1 |
More Information
Set Membership: |
Intragenic Exonic |