Assay Details
Target Gene Details
Entrez Gene ID: | 320806 |
Gene Name: | G elongation factor, mitochondrial 2 |
Gene Aliases: |
6530419G12Rik, A930009M04Rik, EF-G2mt, EFG2, MST027, RRF2mt |
Location: |
Chr.13:97137937-97181196 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gfm2 | NM_001146043.2 | NP_001139515.1 | ||
NM_001271463.1 | 21 | 2590 | NP_001258392.1 | |
NM_001271464.1 | 20 | 2509 | NP_001258393.1 | |
NM_001271465.1 | 21 | 2617 | NP_001258394.1 | |
NM_177266.5 | 21 | 2623 | NP_796240.3 | |
AK035819.1 | 21 | 2528 | ||
AK044371.1 | 20 | 2510 | BAC31889.1 | |
AK052633.1 | 5 | 933 | ||
AK083441.1 | 20 | 2629 | ||
AK161797.1 | BAE36579.1 | |||
BC027341.1 | 21 | 2574 | AAH27341.1 | |
BC138233.1 | 21 | 2521 | ||
BC145208.1 | 21 | 2521 | ||
BF023352.1 |
More Information
Set Membership: |
Intragenic Exonic |