Assay Details
Target Gene Details
Entrez Gene ID: | 108760 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 16 |
Gene Aliases: |
5730405L21Rik, AI415388, Galntl1, mpp-GalNAc-T16 |
Location: |
Chr.12:80518313-80603896 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Galnt16 | NM_001081421.1 | 15 | 2819 | NP_001074890.1 |
AB045325.1 | 15 | 2781 | BAA97985.1 | |
AK077419.1 | 1 | 559 | ||
AK138883.1 | 15 | 2774 | BAE23810.1 | |
AK173105.1 | 16 | 2931 | BAD32383.1 |
More Information
Set Membership: |
Intragenic Exonic |