Assay Details
Target Gene Details
Entrez Gene ID: | 211151 |
Gene Name: | churchill domain containing 1 |
Gene Aliases: |
1700030P15Rik, My015, chch |
Location: |
Chr.12:76765573-76783180 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Churc1 | NM_206534.1 | 4 | 331 | NP_996257.1 |
BC019472.1 | 3 | 258 | AAH19472.1 | |
BC076499.1 | 4 | 331 | AAH76499.1 |