Assay Details
Target Gene Details
Entrez Gene ID: | 56043 |
Gene Name: | aldo-keto reductase family 1, member E1 |
Gene Aliases: |
1810061I10Rik, Akr1e2 |
Location: |
Chr.13:4592490-4609164 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Akr1e1 | NM_018859.2 | 10 | 1179 | NP_061347.2 |
AK002507.1 | 10 | 1154 | BAB22152.1 | |
AK007924.1 | 10 | 1155 | ||
BC012692.1 | 10 | 1195 | AAH12692.1 | |
BQ174229.1 | 1 | 551 | ||
U68535.1 | 10 | 1165 | AAB37274.1 |
More Information
Set Membership: |
Intragenic Exonic |