Assay Details
Target Gene Details
Entrez Gene ID: | 71226 |
Gene Name: | RIKEN cDNA 4933433G19 gene |
Gene Aliases: |
- |
Location: |
Chr.13:66996818-67032405 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4933433G19Rik | NR_045851.1 | |||
AK017041.1 |
Target Gene Details
Entrez Gene ID: | 19210 |
Gene Name: | phosphatidylserine synthase 1 |
Gene Aliases: |
AU044268, AW539008, PSS-1, mKIAA0024 |
Location: |
Chr.13:66932830-66998401 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptdss1 | NM_008959.3 | 13 | 2469 | NP_032985.2 |
AF042731.1 | 13 | 1884 | AAB97845.1 | |
AK036990.1 | 13 | 1931 | BAC29660.1 | |
AK054101.1 | 13 | 1908 | BAC35656.1 | |
AK085857.1 | 13 | 2470 | BAC39555.1 | |
AK086136.1 | 13 | 2128 | ||
AK088048.1 | 13 | 1951 | BAC40117.1 | |
AK137681.1 | 13 | 2139 | BAE23459.1 | |
AK172873.1 | 12 | 4255 | BAD32151.1 | |
AK184832.1 | 1 | 174 | ||
BC003260.1 | 13 | 2106 | AAH03260.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |