Assay Details
Target Gene Details
Entrez Gene ID: | 79221 |
Gene Name: | histone deacetylase 9 |
Gene Aliases: |
AV022454, D030072B18Rik, HD7B, HD9, HDRP, Hdac7b, Mitr, mKIAA0744 |
Location: |
Chr.12:34047580-34917095 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac9 | NM_001271386.1 | NP_001258315.1 | ||
NM_024124.3 | 12 | 2675 | NP_077038.2 | |
AK133334.1 | ||||
AK144242.1 | 3 | 1150 | BAE25793.1 | |
AK173017.1 | BAD32295.1 | |||
BC098187.1 | 12 | 2660 | AAH98187.1 |
More Information
Set Membership: |
Intragenic Exonic |