Assay Details
Target Gene Details
Entrez Gene ID: | 66082 |
Gene Name: | abhydrolase domain containing 6 |
Gene Aliases: |
0610041D24Rik, AA673485, AV065425 |
Location: |
Chr.14:8002807-8056773 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abhd6 | NM_025341.3 | 9 | 1833 | NP_079617.2 |
AK090076.1 | 9 | 1833 | BAC41081.1 | |
AK168782.1 | 8 | 1665 | BAE40616.1 |
More Information
Set Membership: |
Intragenic Exonic |