Assay Details
Target Gene Details
Entrez Gene ID: | 19720 |
Gene Name: | tripartite motif-containing 27 |
Gene Aliases: |
AW538890, Gm19403, Rfp |
Location: |
Chr.13:21179440-21194723 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trim27 | NM_009054.3 | 8 | 1650 | NP_033080.2 |
AK031744.1 | 8 | 1368 | ||
AK088180.1 | 8 | 1134 | BAC40192.1 | |
AK207897.1 | 3 | 200 | ||
BC003219.1 | 8 | 1155 | AAH03219.1 | |
BC069924.1 | 8 | 1361 | AAH69924.1 | |
X75343.1 | 8 | 1065 | CAA53092.1 |
More Information
Set Membership: |
Intragenic Exonic |