Assay Details
Target Gene Details
Entrez Gene ID: | 75512 |
Gene Name: | glutathione peroxidase 6 |
Gene Aliases: |
1700020G18Rik, Ry2d1, olfactory |
Location: |
Chr.13:21295368-21319625 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpx6 | NM_145451.3 | 5 | 1008 | NP_663426.2 |
AA068203.1 | ||||
AK006161.1 | ||||
AV001252.1 | 1 | 192 | ||
AV001457.1 | 1 | 131 | ||
BC013526.1 | 5 | 985 | AAH13526.1 |
More Information
Set Membership: |
Intragenic Exonic |