Assay Details
Target Gene Details
Entrez Gene ID: | 14238 |
Gene Name: | forkhead box F2 |
Gene Aliases: |
FREAC2, Fkh20, LUN |
Location: |
Chr.13:31625816-31631406 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxf2 | NM_010225.2 | 2 | 1905 | NP_034355.2 |
AK132385.1 | 2 | 1906 | BAE21137.1 | |
BC137947.1 | 2 | 1817 | ||
BC137949.1 | 2 | 1817 | ||
Y11148.1 | 2 | 1612 | CAA72035.1 |