Assay Details
Target Gene Details
Entrez Gene ID: | 23876 |
Gene Name: | fibulin 5 |
Gene Aliases: |
A55, DANCE, EVEC |
Location: |
Chr.12:101746565-101819130 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fbln5 | NM_011812.4 | 1 | 336 | NP_035942.1 |
AF112151.1 | 1 | 286 | AAD41767.1 | |
AK084895.1 | 1 | 177 | ||
AK085170.1 | 1 | 166 | BAC39381.1 | |
AK090129.1 | 1 | 275 | BAC41109.1 | |
AK157936.1 | 1 | 288 | ||
AK159189.1 | 1 | 99 | ||
AK159471.1 | 1 | 319 | BAE35112.1 | |
AK159960.1 | 1 | 306 | BAE35515.1 | |
AK165018.1 | 1 | 129 | BAE38002.1 | |
AK178699.1 | ||||
AK208765.1 | ||||
AK219368.1 | ||||
BC006636.1 | AAH06636.1 | |||
BQ557120.1 | 1 | 336 |
More Information
Set Membership: |
Intragenic Exonic |