Assay Details
Target Gene Details
Entrez Gene ID: | 67272 |
Gene Name: | CKLF-like MARVEL transmembrane domain containing 5 |
Gene Aliases: |
1500005P16Rik, 2900052H21Rik, Cklfsf5 |
Location: |
Chr.14:54936470-54939277 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cmtm5 | NM_026066.2 | 1 | 150 | NP_080342.1 |
NM_029390.1 | 1 | 123 | NP_083666.1 | |
AK005168.1 | 1 | 157 | BAB23858.1 | |
AK005375.1 | 1 | 105 | ||
AK013666.1 | 1 | 150 | BAB28947.1 | |
AY241867.1 | 1 | 140 | AAP33489.1 | |
BC049665.1 | 1 | 137 | AAH49665.1 | |
DQ115983.1 | 1 | 123 | AAZ20316.1 |
More Information
Set Membership: |
Intragenic Exonic |