Assay Details
Target Gene Details
Entrez Gene ID: | 12759 |
Gene Name: | clusterin |
Gene Aliases: |
AI893575, ApoJ, Cli, D14Ucla3, SP-40, Sgp-2, Sgp2, Sugp-2 |
Location: |
Chr.14:65968483-65981547 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clu | NM_013492.2 | 5 | 706 | NP_038520.2 |
AF248057.1 | 5 | 589 | AAF67184.1 | |
AF248058.1 | 2 | 260 | AAF67185.1 | |
BC075668.1 | 5 | 535 | AAH75668.1 | |
D14077.1 | 5 | 564 | BAA03162.1 | |
L05670.1 | 5 | 564 | AAA37284.1 | |
L08235.1 | 4 | 536 | AAA37422.1 | |
S70244.1 | 4 | 500 | AAB30623.1 |