Assay Details
Target Gene Details
Entrez Gene ID: | 54616 |
Gene Name: | exostoses (multiple)-like 3 |
Gene Aliases: |
2900009G18Rik, AU040105, Ext1l, mKIAA0519 |
Location: |
Chr.14:65052059-65149899 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Extl3 | NM_018788.3 | 7 | 5059 | NP_061258.2 |
AK137842.1 | 5 | 4136 | BAE23506.1 | |
BC065073.1 | 5 | 4524 | AAH65073.1 |
More Information
Set Membership: |
Intragenic Exonic |