Assay Details
Target Gene Details
Entrez Gene ID: | 26448 |
Gene Name: | MOK protein kinase |
Gene Aliases: |
RAGE1, Rage, Stk30 |
Location: |
Chr.12:110807798-110840957 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mok | NM_011973.2 | 12 | 1421 | NP_036103.1 |
AB022695.1 | 12 | 1421 | BAA81689.1 | |
BC037614.1 | 6 | 2547 | AAH37614.1 | |
BC087925.1 | 7 | 860 | AAH87925.1 |
More Information
Set Membership: |
Intragenic Exonic |