Assay Details
Target Gene Details
Entrez Gene ID: | 18740 |
Gene Name: | paired-like homeodomain transcription factor 1 |
Gene Aliases: |
Bft, P-OTX, Potx, Ptx1 |
Location: |
Chr.13:55825046-55836194 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pitx1 | NM_011097.2 | 1 | 217 | NP_035227.1 |
AK142455.1 | BAE25071.1 | |||
BC012696.1 | AAH12696.1 | |||
U71206.1 | 1 | 217 | AAB16860.1 |
More Information
Set Membership: |
Intragenic Exonic |