Assay Details
Target Gene Details
Entrez Gene ID: | 23849 |
Gene Name: | Kruppel-like factor 6 |
Gene Aliases: |
AI448727, BCD1, C86813, CPBP, Copeb, FM2, FM6, Ierepo1, Ierepo3, R75280, Zf9 |
Location: |
Chr.13:5861489-5870393 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klf6 | NM_011803.2 | 1 | 236 | NP_035933.2 |
AF072403.1 | 1 | 53 | AAC99478.1 | |
AK053584.1 | 1 | 209 | BAC35438.1 | |
AK144137.1 | 1 | 60 | BAE25722.1 | |
AK146615.1 | 1 | 187 | BAE27304.1 | |
AK147904.1 | 1 | 213 | BAE28217.1 | |
AK151769.1 | 1 | 58 | BAE30675.1 | |
AK159420.1 | 1 | 94 | BAE35068.1 | |
AY027436.1 | 1 | 231 | AAK11733.1 | |
BC020042.1 | 1 | 179 | ||
BY128594.1 | 1 | 236 |
More Information
Set Membership: |
Intragenic Exonic |