Assay Details
Target Gene Details
Entrez Gene ID: | 13644 |
Gene Name: | embryonal Fyn-associated substrate |
Gene Aliases: |
- |
Location: |
Chr.14:54916543-54926788 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Efs | NM_010112.4 | 4 | 2133 | NP_034242.2 |
AK030321.1 | 4 | 1260 | BAC26900.1 | |
AK163191.1 | 4 | 1174 | BAE37228.1 | |
BC005438.1 | 4 | 1202 | AAH05438.1 | |
BC031543.1 | 4 | 928 | AAH31543.1 | |
U28728.1 | 4 | 1157 | AAC52340.1 | |
U57686.1 | 4 | 857 | AAB02246.1 |