Assay Details
Target Gene Details
Entrez Gene ID: | 16981 |
Gene Name: | leucine rich repeat protein 3, neuronal |
Gene Aliases: |
NLRR-3 |
Location: |
Chr.12:41451668-41486057 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrrn3 | NM_001271708.1 | 3 | 1029 | NP_001258637.1 |
NM_001271709.1 | 2 | 995 | NP_001258638.1 | |
NM_010733.3 | 2 | 1000 | NP_034863.1 | |
AK036316.1 | 2 | 897 | BAC29381.1 | |
BC069041.1 | 2 | 694 | AAH69041.1 | |
D49802.1 | 2 | 911 | BAA08622.1 |
More Information
Set Membership: |
Intragenic Exonic |