Assay Details
Target Gene Details
Entrez Gene ID: | 70405 |
Gene Name: | calmodulin-like 3 |
Gene Aliases: |
2310068O22Rik, AI326174 |
Location: |
Chr.13:3802893-3804318 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Calml3 | NM_027416.3 | 1 | 1191 | NP_081692.1 |
AK010118.1 | 1 | 1189 | BAB26712.1 | |
BB144976.1 | 1 | 73 | ||
BC005457.1 | 1 | 1186 | AAH05457.1 | |
BX516975.1 | 1 | 367 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)