Assay Details
Target Gene Details
Entrez Gene ID: | 66336 |
Gene Name: | centromere protein P |
Gene Aliases: |
1700022C02Rik, 4921518G09Rik |
Location: |
Chr.13:49464025-49652879 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cenpp | NM_025495.3 | NP_079771.1 | ||
AK006234.1 | BAB24472.1 | |||
AK012863.1 | BAB28519.1 | |||
AK014919.1 | BAB29620.1 | |||
AK076577.1 | BAC36401.1 | |||
BC147207.1 | ||||
BC147208.1 |
Target Gene Details
Entrez Gene ID: | 27047 |
Gene Name: | osteomodulin |
Gene Aliases: |
OSAD, SLRR2C |
Location: |
Chr.13:49579754-49592612 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Omd | NM_012050.2 | 4 | 1479 | NP_036180.1 |
AB007848.1 | 4 | 1484 | BAA22790.1 | |
AK050297.1 | 2 | 582 | BAC34172.1 | |
BC095997.1 | 3 | 1285 | AAH95997.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |