Assay Details
Target Gene Details
Entrez Gene ID: | 16392 |
Gene Name: | ISL1 transcription factor, LIM/homeodomain |
Gene Aliases: |
- |
Location: |
Chr.13:116298257-116309721 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Isl1 | NM_021459.4 | 1 | 201 | NP_067434.3 |
AJ132765.1 | 1 | 67 | CAB38446.1 | |
AK044406.1 | 1 | 188 | ||
AK090263.1 | 1 | 202 | BAC41153.1 | |
AK143052.1 | 1 | 200 | ||
BC132263.1 | 1 | 127 | ||
BC132609.1 | 1 | 127 |