Assay Details
Target Gene Details
Entrez Gene ID: | 75302 |
Gene Name: | additional sex combs like 2 (Drosophila) |
Gene Aliases: |
4930556B16Rik, mKIAA1685 |
Location: |
Chr.12:3426857-3506852 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Asxl2 | NM_001270988.1 | 11 | 1923 | NP_001257917.1 |
NM_172421.5 | 10 | 1779 | NP_766009.2 | |
AK035679.1 | 5 | 2351 | ||
AK036839.1 | 10 | 1736 | BAC29602.1 | |
AK122540.1 | 2 | 552 | BAC65822.1 | |
AK148015.1 | 10 | 1784 | ||
AK162455.1 | 2 | 520 | BAE36927.1 | |
BC141140.1 | 11 | 1757 |