Assay Details
Target Gene Details
Entrez Gene ID: | 238384 |
Gene Name: | solute carrier family 24 (sodium/potassium/calcium exchanger), member 4 |
Gene Aliases: |
A930002M03Rik, AW492432, NCKX4 |
Location: |
Chr.12:102128652-102269083 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc24a4 | NM_172152.2 | 17 | 1748 | NP_742164.1 |
AK044239.1 | 16 | 3782 | BAC31835.1 | |
AK199037.1 | ||||
AY156046.1 | 17 | 1748 | AAN37415.1 | |
BC137945.1 | 17 | 1719 | ||
BC171939.1 | 17 | 1719 |
More Information
Set Membership: |
Intragenic Exonic |