Assay Details
Target Gene Details
Entrez Gene ID: | 11855 |
Gene Name: | Rho GTPase activating protein 5 |
Gene Aliases: |
p190-B, p190B |
Location: |
Chr.12:52503771-52571978 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arhgap5 | NM_009706.2 | 1 | 3238 | NP_033836.2 |
AK041016.1 | 2 | 3270 | ||
AK131622.1 | BAE20727.1 | |||
BC150823.1 | 1 | 3208 |
More Information
Set Membership: |
Intragenic Exonic |