Assay Details
Target Gene Details
Entrez Gene ID: | 26363 |
Gene Name: | biotinidase |
Gene Aliases: |
- |
Location: |
Chr.14:31640999-31668197 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Btd | NM_025295.4 | 2 | 159 | NP_079571.1 |
AK005506.1 | 2 | 152 | BAB24086.1 | |
AK135375.1 | 2 | 149 | BAE22509.1 | |
BC024051.1 | 2 | 126 | ||
CJ181966.1 | 2 | 159 |
More Information
Set Membership: |
Intragenic Exonic |