Assay Details
Target Gene Details
Entrez Gene ID: | 18260 |
Gene Name: | occludin |
Gene Aliases: |
AI503564, Ocl |
Location: |
Chr.13:100496504-100553689 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ocln | NM_008756.2 | 3 | 573 | NP_032782.1 |
AK019880.1 | 3 | 525 | BAB31900.1 | |
AK033532.1 | 3 | 506 | ||
AK042576.1 | 3 | 573 | BAC31298.1 | |
AK145798.1 | BAE26656.1 | |||
BC138679.1 | 3 | 556 | ||
BC138680.1 | 3 | 556 | ||
BC145371.1 | 3 | 571 | ||
U49185.1 | 3 | 616 | AAC52515.1 |
More Information
Set Membership: |
Intragenic Exonic |