Assay Details
Target Gene Details
Entrez Gene ID: | 15487 |
Gene Name: | hydroxysteroid (17-beta) dehydrogenase 3 |
Gene Aliases: |
- |
Location: |
Chr.13:64058274-64089258 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hsd17b3 | NM_008291.3 | 11 | 947 | NP_032317.2 |
BB041573.2 | 3 | 328 | ||
U66827.1 | 11 | 932 | AAB06793.1 |