Assay Details
Target Gene Details
Entrez Gene ID: | 69706 |
Gene Name: | leucine rich repeat protein 1 |
Gene Aliases: |
2410005L11Rik, AU021780, LRR-1, Ppil5 |
Location: |
Chr.12:69168814-69179010 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrr1 | NM_001081406.1 | 4 | 1212 | NP_001074875.1 |
AK010419.1 | 3 | 483 | BAB26922.1 | |
AK144741.1 | 4 | 1213 | ||
BC022648.1 | 3 | 444 | AAH22648.1 | |
BC117053.1 | 3 | 469 | AAI17054.1 | |
BC117055.1 | 3 | 469 | AAI17056.1 |
More Information
Set Membership: |
Intragenic Exonic |