Assay Details
Target Gene Details
Entrez Gene ID: | 544971 |
Gene Name: | B double prime 1, subunit of RNA polymerase III transcription initiation factor IIIB |
Gene Aliases: |
AI662220, AW049390, AW456745, B130055N23Rik, G630013P12Rik, TAF3B1, TFC5, TFIIIB150, TFIIIB90, Tfnr, mKIAA1241 |
Location: |
Chr.13:100017994-100104073 on Build GRCm38 |
Assay Gene Location: | Within Exon 37 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bdp1 | NM_001081061.1 | 37 | 7033 | NP_001074530.1 |
AK078583.1 | 5 | 520 | ||
AK090184.1 | 6 | 663 | BAC41127.1 | |
AK220262.1 | 27 | 5281 | BAD90187.1 | |
BC030359.1 | 9 | 1256 | AAH30359.1 | |
BC144852.1 | 18 | 2723 |
More Information
Set Membership: |
Intragenic Exonic |