Assay Details
Target Gene Details
Entrez Gene ID: | 13865 |
Gene Name: | nuclear receptor subfamily 2, group F, member 1 |
Gene Aliases: |
COUP-TF1, COUP-TFI, COUPTFA, EAR3, Erbal3, SVP44, Tcfcoup1 |
Location: |
Chr.13:78188971-78199172 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nr2f1 | NM_010151.2 | 2 | 1125 | NP_034281.2 |
AK014008.1 | 1 | 214 | ||
AK148880.1 | 2 | 1462 | BAE28687.1 | |
BC108408.1 | 2 | 1014 | AAI08409.1 | |
U07625.1 | 2 | 727 | AAA19853.1 | |
X74134.1 | 2 | 1025 | CAA52231.1 |
More Information
Set Membership: |
Intragenic Exonic |