Assay Details
Target Gene Details
Entrez Gene ID: | 27260 |
Gene Name: | pleckstrin 2 |
Gene Aliases: |
- |
Location: |
Chr.12:78888691-78907182 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plek2 | NM_013738.3 | 9 | 1040 | NP_038766.1 |
AF157600.1 | 9 | 1086 | AAD42973.1 | |
AF170564.1 | 9 | 1052 | AAD46924.1 | |
BC028902.1 | 9 | 1040 | AAH28902.1 |
More Information
Set Membership: |
Intragenic Exonic |