Assay Details
Target Gene Details
Entrez Gene ID: | 192119 |
Gene Name: | dicer 1, ribonuclease type III |
Gene Aliases: |
1110006F08Rik, D12Ertd7e, mKIAA0928 |
Location: |
Chr.12:104687740-104751952 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dicer1 | NM_148948.2 | 24 | 5417 | NP_683750.2 |
AB081470.1 | 23 | 5215 | BAC15765.1 | |
AF430845.1 | 24 | 5425 | AAM21495.1 | |
AK129241.1 | 18 | 3976 | BAC98051.1 | |
AK137929.1 | 3 | 1811 | BAE23515.1 |