Assay Details
Target Gene Details
Entrez Gene ID: | 59079 |
Gene Name: | Erbb2 interacting protein |
Gene Aliases: |
1700028E05Rik, Erbin, mKIAA1225 |
Location: |
Chr.13:103818786-103920586 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Erbb2ip | NM_001005868.2 | 27 | 5998 | NP_001005868.1 |
NM_001289473.1 | 26 | 5854 | NP_001276402.1 | |
NM_001289474.1 | 24 | 5530 | NP_001276403.1 | |
NM_001289475.1 | 26 | 5881 | NP_001276404.1 | |
AK051733.1 | 6 | 2513 | BAC34742.1 | |
AK122473.1 | 26 | 5455 | BAC65755.1 | |
BC005691.1 | 6 | 1979 | ||
BC028256.1 | 7 | 3441 | AAH28256.1 |
More Information
Set Membership: |
Intragenic Exonic |