Assay Details
Target Gene Details
Entrez Gene ID: | 20787 |
Gene Name: | sterol regulatory element binding transcription factor 1 |
Gene Aliases: |
ADD-1, ADD1, D630008H06, SREBP-1, SREBP-1a, SREBP-1c, SREBP1, SREBP1c, bHLHd1 |
Location: |
Chr.11:60199089-60222686 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Srebf1 | NM_011480.3 | 19 | 3944 | NP_035610.1 |
AK040663.1 | 1 | 302 | ||
AK052628.1 | 19 | 3945 | BAC35068.1 | |
BC056922.1 | 19 | 3926 | AAH56922.1 |