Assay Details
Target Gene Details
Entrez Gene ID: | 78887 |
Gene Name: | Sfi1 homolog, spindle assembly associated (yeast) |
Gene Aliases: |
- |
Location: |
Chr.11:3131850-3203343 on Build GRCm38 |
Assay Gene Location: | Within Exon 33 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sfi1 | NM_030207.2 | 33 | 3888 | NP_084483.2 |
AK019095.1 | 10 | 2872 | BAB31543.1 | |
AK046135.1 | 22 | 2401 | ||
AK046373.1 | 19 | 2403 | ||
AK129159.1 | 13 | 3170 | BAC97969.1 | |
AK133558.1 | 33 | 3836 | BAE21725.1 | |
AK181569.1 | 3 | 291 | ||
BC026390.1 | 22 | 2529 | AAH26390.1 | |
BC046305.1 | 25 | 3608 | AAH46305.1 | |
BC048950.1 | 16 | 1916 | AAH48950.1 |