Assay Details
Target Gene Details
Entrez Gene ID: | 237987 |
Gene Name: | otopetrin 2 |
Gene Aliases: |
4732464P15 |
Location: |
Chr.11:115307163-115332306 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Otop2 | NM_172801.2 | 3 | 528 | NP_766389.2 |
AB087504.1 | 2 | 413 | BAC67684.1 | |
AK028866.1 | 3 | 529 | BAC26159.1 |
More Information
Set Membership: |
Intragenic Exonic |