Assay Details
Target Gene Details
Entrez Gene ID: | 66176 |
Gene Name: | N-acetyltransferase 9 (GCN5-related, putative) |
Gene Aliases: |
1110028N05Rik |
Location: |
Chr.11:115182831-115187886 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nat9 | NM_025400.3 | 7 | 990 | NP_079676.1 |
AK003948.1 | 7 | 944 | BAB23087.1 | |
AK009262.1 | 6 | 720 | ||
AK020083.1 | 7 | 973 | BAC25613.1 | |
AK082469.1 | 7 | 930 | BAC38501.1 | |
AK148048.1 | 7 | 1022 | BAE28311.1 | |
AK154593.1 | 6 | 1121 | ||
AK213540.1 | 1 | 226 | ||
AK215096.1 | 1 | 234 | ||
BC011315.1 | 7 | 990 | AAH11315.1 |