Assay Details
Target Gene Details
Entrez Gene ID: | 216810 |
Gene Name: | target of myb1-like 2 (chicken) |
Gene Aliases: |
2900016I08Rik, A730055F12Rik, AU042072, Srebf1 |
Location: |
Chr.11:60223796-60352908 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tom1l2 | NM_001039092.3 | 14 | 2007 | NP_001034181.1 |
NM_153080.2 | 15 | 2067 | NP_694720.2 | |
BC062947.1 | 14 | 1971 | ||
BC127266.1 | 14 | 1973 |