Assay Details
Target Gene Details
Entrez Gene ID: | 210004 |
Gene Name: | UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase-like 1 |
Gene Aliases: |
6030413G23Rik |
Location: |
Chr.11:121616197-121673160 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
B3gntl1 | NM_178664.5 | 13 | 1754 | NP_848779.2 |
AK031361.1 | 13 | 1743 | BAC27366.1 | |
AK156318.1 | 13 | 1732 | BAE33671.1 | |
BC022739.1 | 7 | 1066 | AAH22739.1 | |
BC088993.1 | 13 | 1619 | AAH88993.1 |
More Information
Set Membership: |
Intragenic Exonic |