Assay Details
Target Gene Details
Entrez Gene ID: | 11302 |
Gene Name: | apoptosis-associated tyrosine kinase |
Gene Aliases: |
AATYK, aatyk1, mKIAA0641 |
Location: |
Chr.11:120007311-120047182 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aatk | NM_001198785.1 | 11 | 2639 | NP_001185714.1 |
NM_001198787.1 | 11 | 2411 | NP_001185716.1 | |
NM_007377.4 | 11 | 2922 | NP_031403.2 | |
AB093253.2 | 11 | 2922 | BAC41437.2 | |
AB288871.1 | 11 | 2541 | ||
AF011908.1 | 11 | 2378 | AAB71837.1 | |
AK036705.1 | 11 | 2413 | BAC29541.1 | |
AK147239.1 | 8 | 2432 | BAE27789.1 | |
AK155100.1 | 11 | 2413 | BAE33045.1 | |
AY236858.1 | 11 | 2417 | AAO92350.1 | |
AY236859.1 | 11 | 2444 | AAO92351.1 | |
BC080846.1 | 11 | 2493 | AAH80846.1 |
More Information
Set Membership: |
Intragenic Exonic |