Assay Details
Target Gene Details
Entrez Gene ID: | 217057 |
Gene Name: | peptidyl-tRNA hydrolase 2 |
Gene Aliases: |
A230072I16Rik, Bit1, CGI-147 |
Location: |
Chr.11:86683983-86692457 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptrh2 | NM_001098810.2 | 2 | 2446 | NP_001092280.1 |
NM_175004.2 | 3 | 2637 | NP_778169.1 | |
AK028555.1 | 3 | 2637 | BAC26006.1 | |
AK031620.1 | 2 | 2448 | BAC27482.1 | |
AK038888.1 | 2 | 2492 | BAC30159.1 | |
AK086996.1 | 3 | 2574 |