Assay Details
Target Gene Details
Entrez Gene ID: | 103554 |
Gene Name: | proteasome (prosome, macropain) activator subunit 4 |
Gene Aliases: |
AA409398, AU041366, TEMO, mKIAA0077 |
Location: |
Chr.11:30771328-30881150 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psme4 | NM_134013.3 | 10 | 1422 | NP_598774.2 |
AF296169.1 | 10 | 1133 | AAG09060.1 | |
AK052279.1 | 1 | 2465 | ||
BC141381.1 | 10 | 1135 | ||
CV557303.1 | 3 | 291 |