Assay Details
Target Gene Details
Entrez Gene ID: | 380732 |
Gene Name: | mast cell immunoglobulin like receptor 1 |
Gene Aliases: |
Allergin-1, Gm885, Mca32 |
Location: |
Chr.11:106750652-106768794 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Milr1 | NM_001033435.3 | 3 | 346 | NP_001028607.1 |
NM_001271372.1 | 3 | 346 | NP_001258301.1 | |
NM_001271373.1 | 3 | 346 | NP_001258302.1 | |
NM_001271374.1 | 3 | 346 | NP_001258303.1 | |
NM_001271375.1 | 3 | 346 | NP_001258304.1 | |
NR_073165.1 | 3 | 346 | ||
AB542953.1 | 3 | 304 | ||
AK136488.1 | 3 | 306 | BAE23003.1 | |
AK137500.1 | 3 | 305 | BAE23382.1 | |
AK151814.1 | 3 | 304 | BAE30712.1 | |
AK153506.1 | 3 | 254 | BAE32051.1 | |
AK169925.1 | 3 | 309 | BAE41463.1 | |
AK170947.1 | 3 | 305 | BAE42134.1 | |
AK171381.1 | 3 | 305 | BAE42422.1 | |
BC139224.1 | 3 | 332 | ||
BC139225.1 | 3 | 332 | ||
BC145580.1 | 3 | 332 | ||
BC145582.1 | 3 | 332 | ||
BY197245.1 |
Target Gene Details
Entrez Gene ID: | 50776 |
Gene Name: | polymerase (DNA directed), gamma 2, accessory subunit |
Gene Aliases: |
- |
Location: |
Chr.11:106754821-106780010 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polg2 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |