Assay Details
Target Gene Details
Entrez Gene ID: | 20305 |
Gene Name: | chemokine (C-C motif) ligand 6 |
Gene Aliases: |
MRP-1, Scya6, c10 |
Location: |
Chr.11:83587886-83593087 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl6 | NM_009139.3 | 4 | 555 | NP_033165.1 |
AF128188.1 | AAF22529.1 | |||
AF128189.1 | AAF22530.1 | |||
AF128190.1 | AAF22531.1 | |||
AF128191.1 | AAF22532.1 | |||
AF128192.1 | AAF22533.1 | |||
AK002697.1 | 4 | 458 | BAB22291.1 | |
AK008547.1 | 4 | 459 | BAB25734.1 | |
AK150336.1 | 4 | 458 | BAE29478.1 | |
AK154553.1 | 4 | 459 | BAE32672.1 | |
BC002073.1 | 4 | 448 | AAH02073.1 | |
M58004.1 | 4 | 458 | AAA37329.1 |
Target Gene Details
Entrez Gene ID: | 100504464 |
Gene Name: | RIKEN cDNA E230016K23 gene |
Gene Aliases: |
- |
Location: |
Chr.11:83582061-83623693 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
E230016K23Rik | NR_036452.1 | |||
NR_036453.1 | ||||
AK054074.1 | BAC35646.1 | |||
AK087588.1 | ||||
AK154990.1 | BAE32978.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |